missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Tribasic, Anhydrous, Technical, Spectrum™ Chemical

Catalog No. p-7158623
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606718 500 g Poly Bottle
18606719 2.5 kg Poly Bottle
18606720 12 kg Poly Pail
18606721 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606718 Supplier Spectrum Chemical Mfg Cor Supplier No. S1409500GM
Only null left

S1409, 7601-54-9, Na3PO4

Spectrum™ Chemical Sodium Phosphate Tribasic, Anhydrous, Technical or trisodium phosphate, is used as a cleaning agent and is sufficiently alkaline to saponify grease and oils. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.94
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
IUPAC Name trisodium phosphate
SMILES [Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Poly Bottle