missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Tribasic, Anhydrous, FCC, 97%, Spectrum™ Chemical

Catalog No. p-7158260
Change view
Click to view available options
Quantity:
500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606740 500 g Poly Bottle
18606742 12 kg Poly Pail
18606743 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606740 Supplier Spectrum Chemical Mfg Cor Supplier No. S1276500GM
Only null left

S1276, 7601-54-9, Na3PO4

Spectrum™ Chemical Sodium Phosphate, Tribasic, Anhydrous, FCC is used as an acidity regulator (buffering agent), emulsifier, thickening agent, nutrition enlargement agent and sequestrant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.94
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
IUPAC Name trisodium phosphate
SMILES [Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 97%
Grade FCC
Loss on Ignition 0.02
Packaging Poly Bottle