missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate monobasic dihydrate, meets analytical spec. of BP, FCC, Ph. Eur., USP, E 339, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Meets analytical specification of BP, FCC, Ph. Eur., USP, E 339, 98-100.5% (calc. to the dried substance)
Supplier: Honeywell Chemicals 042695KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Phosphate Monobasic Dihydrate | |
| 13472-35-0 | |
| H6NaO6P | |
| MFCD00149209 | |
| sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
| O.O.OP(=O)(O)[O-].[Na+] | |
| 156.006 | |
| 156.01g/mol |
| Meets analytical specification of Ph. Eur., BP, USP, FCC | |
| 5 kg | |
| NaH2PO4 · 2H2O | |
| NONH for all modes of transport | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| sodium;dihydrogen phosphate;dihydrate | |
| 23673460 | |
| 98 to 100.5% (calc. to the dried substance) |
Chemical Identifiers
| 13472-35-0 | |
| 156.006 | |
| VBJGJHBYWREJQD-UHFFFAOYSA-M | |
| 23673460 | |
| O.O.OP(=O)(O)[O-].[Na+] |
| H6NaO6P | |
| MFCD00149209 | |
| sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
| sodium;dihydrogen phosphate;dihydrate |
Safety and Handling
EINECSNumber : 231-449-2
RTECSNumber : WA1900500