missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic dihydrate, 99+%, for analysis

Catalog No. AC271750010
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271750010 1 kg Plastic Bottle
AC271750025 2.5 kg Plastic Bottle
AC271750100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC271750010 Supplier Thermo Scientific Chemicals Supplier No. 271750010
Only null left

CAS: 13472-35-0 | H2NaO42H2O | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H2NaO42H2O
Molecular Weight (g/mol) 156.01
MDL Number MFCD00149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Melting Point 60.0°C
Color White
Physical Form Adhering Crystals
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula NaH2PO4·2H2O
Merck Index 15, 8790
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 156.01
Percent Purity ≥99%
Grade Analytical
Loss on Drying 24% max.
Packaging Plastic Bottle
Chemical Name or Material Sodium phosphate, monobasic dihydrate
Show More Show Less

RUO – Research Use Only