missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, Heptahydrate, USP, EP, Pyrogen Free, 98-102.5%, Macco Organiques

Catalog No. 18614847
Change view
Click to view available options
Quantity:
12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18614847 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614847 Supplier Spectrum Chemical Mfg Cor Supplier No. 51115070120KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Macco products offered by Spectrum™ Chemical are suitable for biopharmaceutical production.

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Color Clear/Colorless
Quantity 12 kg
Percent Purity 98 to 102.5%
Grade EP/USP
Packaging Plastic Pail