missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate, Ultrapure, 500 g, Innovating Science™

Catalog No. S000976
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000976 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000976 Supplier Aldon Corporation Supplier No. IS29046
  • Appearance: White to Off-White Powder
  • Purity: ≥ 99.0%
  • pH (5% Aqueous): 8.7 to 9.3
  • Solubility: Clear and Colorless
  • Identification (FTIR): Conforms to Structure
  • Loss on Drying (4h at 105 °C): 44 to 50%
  • Lead: ≤ 10 ppm
  • Iron: ≤ 10 ppm
  • Chloride: ≤ 10 ppm
  • Phosphate: 35.0% to 39.0%
  • Sulfate: ≤ 50 ppm
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

CAS 7782-85-6
Molecular Weight (g/mol) 268.07

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 100%
Grade Ultra Pure
Packaging HDPE
Chemical Name or Material Sodium phosphate, dibasic, heptahydrate