missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic heptahydrate, Honeywell Fluka™
ACS reagent, 98.0-102.0%
$236.95 - $431.50
Chemical Identifiers
| CAS | 7782-85-6 |
|---|---|
| Molecular Formula | H15Na2O11P |
| Molecular Weight (g/mol) | 268.06 |
| MDL Number | MFCD00149180 |
| InChI Key | LVXHNCUCBXIIPE-UHFFFAOYSA-L |
| Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
| PubChem CID | 6096963 |
| IUPAC Name | disodium phosphoric acid heptahydrate |
| SMILES | O.[Na+].[Na+].OP([O-])([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7782-85-6 | |
| 268.06 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| 6096963 | |
| O.[Na+].[Na+].OP([O-])([O-])=O |
| H15Na2O11P | |
| MFCD00149180 | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| disodium phosphoric acid heptahydrate |
Specifications
| 7782-85-6 | |
| H15Na2O11P | |
| MFCD00149180 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| disodium phosphoric acid heptahydrate | |
| 6096963 | |
| 98 to 102% | |
| Sodium phosphate dibasic heptahydrate |
| 500 g | |
| Na2HPO4 · 7H2O | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| O.[Na+].[Na+].OP([O-])([O-])=O | |
| 268.06 | |
| 268.07g/mol | |
| 1.68g/mL at 25°C; Vapor Density: 4.9 (vs air) |