missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate (Granular/USP), Fisher Chemical

Catalog No. S4713
Change view
Click to view available options
Poly Bottle; 3kg (6.6 lb.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S4713 Poly Bottle; 3kg (6.6 lb.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S4713 Supplier Thermo Fisher Scientific Supplier No. S471 3
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Specifications

Melting Point 48°C
Color White
CAS 7782-85-6
pH 9.0 to 9.3
Physical Form Solid
Quantity 3 kg
Molecular Formula H15Na2O11P
MDL Number MFCD00149180
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
SMILES O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium phosphoric acid heptahydrate
Molecular Weight (g/mol) 268.06
PubChem CID 6096963
Formula Weight 268.07
Percent Purity 98.0 to 100.5% (USP)
Grade USP
Identification Pass Test (USP)
Loss on Drying 43.0 to 50.0% (USP)
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
Show More Show Less