missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, Heptahydrate, Granular, BiotechGrade, 98-100.5%, Spectrum™ Chemical

S1412, 7782-85-6, Na2HPO4·7H2O

Supplier:  Spectrum Chemical Mfg Cor S141225KG

 View more versions of this product

Catalog No. 18606700

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Phosphate Dibasic, Heptahydrate, Granular, BiotechGrade is used along with disodium hydrogen phosphate in food treatment to adjust pH. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Specifications

Specifications

7782-85-6
2.5 kg
MFCD00149180
LVXHNCUCBXIIPE-UHFFFAOYSA-L
disodium phosphoric acid heptahydrate
98.0 to 100.5%
43-50%
Poly Bottle
1
H15Na2O11P
To pass test
O.[Na+].[Na+].OP([O-])([O-])=O
268.06
Biotech
Odorless

Chemical Identifiers

7782-85-6
268.06
LVXHNCUCBXIIPE-UHFFFAOYSA-L
O.[Na+].[Na+].OP([O-])([O-])=O
H15Na2O11P
MFCD00149180
disodium phosphoric acid heptahydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions