missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, Heptahydrate, Granular, BiotechGrade, 98-100.5%, Spectrum™ Chemical

Catalog No. 18606699
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606699 500 g Poly Bottle
18606700 2.5 kg Poly Bottle
18606701 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606699 Supplier Spectrum Chemical Mfg Cor Supplier No. S1412500GM

S1412, 7782-85-6, Na2HPO4·7H2O

Spectrum™ Chemical Sodium Phosphate Dibasic, Heptahydrate, Granular, BiotechGrade is used along with disodium hydrogen phosphate in food treatment to adjust pH. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Solubility Information To pass test
Percent Purity 98.0 to 100.5%
Grade Biotech
Loss on Drying 43-50%
Odor Odorless
Packaging Poly Bottle