Learn More
Sodium Phosphate, Dibasic, Heptahydrate, Granular, ACS, 98-102%, Spectrum™ Chemical
Supplier:  Spectrum Chemical Mfg Cor S140425KG
 View more versions of this product
Catalog No. 18613797
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
7782-85-6
8.7 to 9.3
98 to 102%
Poly Bottle
1
2.5 kg
ACS
Chemical Identifiers
7782-85-6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.