missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Heptahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18613804
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613804 50 kg Poly Bottle
18613800 125 g Poly Bottle
18613801 500 g Poly Bottle
18613802 2.5 kg Poly Bottle
18613803 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613804 Supplier Spectrum Chemical Mfg Cor Supplier No. S140550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7782-85-6

Specifications

pH 8.7 to 9.3
CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Molecular Weight (g/mol) 268.07
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate, Dibasic, Heptahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.