missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate), Reagents

Catalog No. p-200021213
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337699 10 kg
01337698 2.5 kg
01337697 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337699 Supplier Labchem Inc Supplier No. C132400010KG
Only null left

Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate)
CAS 7782-85-6
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Molecular Formula Na2HPO4, 7H2O
Synonym Disodium Hydrogen Phosphate Heptahydrate
Molecular Weight (g/mol) 268.0658
Grade ACS
Packaging Poly Pail
Specific Gravity 1.7
Show More Show Less