missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent

CAS: 7782-85-6 | H15Na2O11P | 268.06 g/mol

$157.16 - $1220.27

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424385000
SDS View Documents
Thermo Scientific Chemicals
424385000
500 g Plastic bottle
Each for $157.16
Only null left
 
AC424380010
SDS View Documents
Thermo Scientific Chemicals
424380010
1 kg Plastic bottle
Each for $277.91
Only null left
 
AC424380050
SDS View Documents
Thermo Scientific Chemicals
424380050
5 kg Plastic Bottle
Each for $1,220.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-85-6
268.06
LVXHNCUCBXIIPE-UHFFFAOYSA-L
6096963
O.[Na+].[Na+].OP([O-])([O-])=O
H15Na2O11P
MFCD00149180
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
disodium phosphoric acid heptahydrate

Specifications

48.0°C
White
Crystals
98+%
Na2HPO4·7H2O
15, 8789
LVXHNCUCBXIIPE-UHFFFAOYSA-L
disodium phosphoric acid heptahydrate
6096963
≥98%
Plastic bottle
1.67
7782-85-6
8.7 to 9.3 (5% soln. at 25°C)
500 g
H15Na2O11P
MFCD00149180
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
O.[Na+].[Na+].OP([O-])([O-])=O
268.06
268.07
ACS Reagent
>48°C
Sodium phosphate, dibasic heptahydrate
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

RTECSNumber : WC4600000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only