missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 48.0°C |
| Color | White |
| pH | 8.7 to 9.3 (5% soln. at 25°C) |
| Physical Form | Crystals |
| Quantity | 500 g |
| Packaging | Plastic bottle |
| Decomposition Information | >48°C |
| Assay Percent Range | 98+% |
| Density | 1.67 |
| Linear Formula | Na2HPO4·7H2O |
| Show More |
Safety and Handling
| Hazard Category | HYGROSCOPIC |
| RTECSNumber | WC4600000 |
RUO – Research Use Only