missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate, 98.0-102.0%, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19144101
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19144101 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19144101 Supplier MP Biomedicals Inc Supplier No. 0219144101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS grade reagent for laboratory use.

Specifications

Color White
CAS 7782-85-6
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.0-102.0%
Molecular Formula H15Na2O11P
MDL Number MFCD00149180
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
SMILES O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium phosphoric acid heptahydrate
Molecular Weight (g/mol) 268.06
PubChem CID 6096963
Formula Weight 268.07
Percent Purity 100%
Grade ACS Reagent
Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
Show More Show Less
Recommended Storage Room Temperature