missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic dihydrate, specified accord. to the requir. of USP, Ph.Eur.
CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol
$627.36 - $627.36
Chemical Identifiers
| CAS | 10028-24-7 |
|---|---|
| Molecular Formula | H5Na2O6P |
| Molecular Weight (g/mol) | 177.99 |
| MDL Number | MFCD00149182 |
| InChI Key | KDQPSPMLNJTZAL-UHFFFAOYSA-L |
| IUPAC Name | disodium dihydrate hydrogen phosphate |
| SMILES | O.O.[Na+].[Na+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC460790050
|
Thermo Scientific Chemicals
460790050 |
5 kg |
Each for $627.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10028-24-7 | |
| 177.99 | |
| KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
| O.O.[Na+].[Na+].OP([O-])([O-])=O |
| H5Na2O6P | |
| MFCD00149182 | |
| disodium dihydrate hydrogen phosphate |
Specifications
| 243.0°C to 245.0°C | |
| 5 kg | |
| Na2HPO4·2 H2O | |
| Solubility aq. soln.: 93 g/L(20°C). | |
| O.O.[Na+].[Na+].OP([O-])([O-])=O | |
| 177.99 | |
| Sodium phosphate dibasic dihydrate |
| 10028-24-7 | |
| H5Na2O6P | |
| MFCD00149182 | |
| KDQPSPMLNJTZAL-UHFFFAOYSA-L | |
| disodium dihydrate hydrogen phosphate | |
| 177.99 |
RUO – Research Use Only