missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Honeywell Fluka™

Catalog No. p-7071719
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001836 1 kg
6001837 2.5 kg
6001838 5 kg
60044547 6 x 1 kg
60044546 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6001836 Supplier Honeywell Chemicals Supplier No. 042721KG
Only null left

Meets analytical specification of BP, Ph. Eur., 98.5-101% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 1 kg
Linear Formula Na2HPO4 · 2H2O
IUPAC Name disodium;hydrogen phosphate;dihydrate
Formula Weight 177.99g/mol
Percent Purity 98.5 to 101% (calc. to the dried substance)
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate, Meets analytical specification of Ph.Eur.BP
EINECSNumber 231-448-7