missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Dihydrate, FCC, 98%, Spectrum™ Chemical

Catalog No. 18606736
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606736 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606736 Supplier Spectrum Chemical Mfg Cor Supplier No. S1108500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1108, 10028-24-7, Na2HPO4·2H2O

Spectrum™ Chemical Sodium Phosphate, Dibasic, Dihydrate, FCC is used in foods to adjust pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Percent Purity 98%
Grade FCC
Loss on Drying 18-22%
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate, Dibasic, Dihydrate