missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Dihydrate, FCC, 98%, Spectrum™ Chemical

Catalog No. 18606737
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606737 2.5 kg Poly Bottle
18606738 12 kg Poly Pail
18606739 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606737 Supplier Spectrum Chemical Mfg Cor Supplier No. S11082.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

S1108, 10028-24-7, Na2HPO4·2H2O

Spectrum™ Chemical Sodium Phosphate, Dibasic, Dihydrate, FCC is used in foods to adjust pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 98%
Grade FCC
Loss on Drying 18-22%
Packaging Poly Bottle