missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic dihydrate, 98%, extra pure

Catalog No. p-7214373
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448150010 1 kg
AC448150050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448150010 Supplier Thermo Scientific Chemicals Supplier No. 448150010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243.0°C to 245.0°C
Quantity 1 kg
Assay Percent Range 98%
Linear Formula Na2HPO4·2 H2O
Merck Index 15, 8789
Solubility Information Solubility aq. soln.: 93 g/L(20°C).
Formula Weight 177.99
Grade Extra Pure
Chemical Name or Material Sodium phosphate, dibasic dihydrate

RUO – Research Use Only