missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, 98+%, for analysis

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol

$75.65 - $218.84

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124992500
SDS View Documents
Thermo Scientific Chemicals
124992500
250 g Glass bottle N/A
Only null left
 
AC124990010
SDS View Documents
Thermo Scientific Chemicals
124990010
1 kg Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10028-24-7
177.99
KDQPSPMLNJTZAL-UHFFFAOYSA-L
6451167
O.O.[Na+].[Na+].OP([O-])([O-])=O
H5Na2O6P
MFCD00149182
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
disodium;hydrogen phosphate;dihydrate

Specifications

243.0°C to 245.0°C
250 g
H5Na2O6P
MFCD00149182
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
KDQPSPMLNJTZAL-UHFFFAOYSA-L
disodium;hydrogen phosphate;dihydrate
6451167
≥98%
Glass bottle
10028-24-7
98+%
Na2HPO4·2H2O
15,8789
(5% in water) Clear colorless
O.O.[Na+].[Na+].OP([O-])([O-])=O
177.99
177.99
Analytical
Sodium phosphate dibasic dihydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only