missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium phosphate, 0.5M buffer soln., pH 7.6

Catalog No. AAJ60158AP
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60158AP 500mL
AAJ60158AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60158AP Supplier Thermo Scientific™ Supplier No. J60158AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.5M buffer soln.
pH 7.6
Quantity 500mL
MDL Number MFCD00146206

RUO – Research Use Only