missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium phosphate, 0.2M buffer soln., pH 8.0

Supplier:  Thermo Scientific™ J62733K2

Catalog No. AAJ62733K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Phosphate buffer is a multi-purpose buffer used for many molecular biology procedures. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Fully miscible in water.

Notes
Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Sodium phosphate buffer soln.
8.0
MFCD00146206
0.2M buffer soln.
1L
Fully miscible in water.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only