missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate (White Crystalline Solid/Certified), Fisher Chemical™

Catalog No. p-34086
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O6114500 500 g Poly Bottle
O61141 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O6114500 Supplier Thermo Fisher Scientific Supplier No. O6114500

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
MDL Number MFCD00003501
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 100°C
Color White
pH 5 to 7
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥98.0 %
Formula Weight 238.09
Percent Purity ≥98.0%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Density 2.6g/cm³
Chemical Name or Material Sodium Persulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause sensitization by inhalation and skin contact.
DOTInformation DOT Class 5.1, : Oxidizer