missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate (Sodium Peroxydisulfate), Reagents

Catalog No. 01336995
Change view
Click to view available options
Quantity:
500 g
12 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336995 12 kg
01336992 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336995 Supplier Labchem Inc Supplier No. C232310012KG
Only null left

Sodium Persulfate (Sodium Peroxydisulfate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.09
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym Sodium Peroxydisulfate
IUPAC Name disodium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade Reagent
Packaging Poly Pail
Specific Gravity 2.6
Chemical Name or Material Sodium Persulfate (Sodium Peroxydisulfate)