missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Persulfate (Sodium Peroxydisulfate), Reagents
Sodium Persulfate (Sodium Peroxydisulfate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2323100225A5
Specifications
| Sodium Persulfate (Sodium Peroxydisulfate) | |
| Colorless/White | |
| Solid | |
| Na2O8S2 | |
| CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
| disodium [(sulfonatoperoxy)sulfonyl]oxidanide | |
| Reagent | |
| 2.6 |
| 120°C | |
| 7775-27-1 | |
| 2.25 kg | |
| Sodium Peroxydisulfate | |
| [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
| 238.09 | |
| Poly Bottle |
Chemical Identifiers
| 7775-27-1 | |
| 238.09 | |
| Sodium Peroxydisulfate | |
| [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| Na2O8S2 | |
| CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
| disodium [(sulfonatoperoxy)sulfonyl]oxidanide |