missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium persulfate, Honeywell
Purum p.a.,≥99.0% (RT)
$172.55 - $172.55
Chemical Identifiers
| CAS | 7775-27-1 |
|---|---|
| Molecular Formula | Na2O8S2 |
| Molecular Weight (g/mol) | 238.092 |
| MDL Number | MFCD00003501 |
| InChI Key | CHQMHPLRPQMAMX-UHFFFAOYSA-L |
| Synonym | sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate |
| PubChem CID | 62655 |
| IUPAC Name | disodium;sulfonatooxy sulfate |
| SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
| 7775-27-1 | |
| 238.092 | |
| CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
| 62655 | |
| [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
| Na2O8S2 | |
| MFCD00003501 | |
| sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
| disodium;sulfonatooxy sulfate |
Specifications
| 7775-27-1 | |
| Na2O8S2 | |
| MFCD00003501 | |
| sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
| [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] | |
| 238.092 | |
| 238.1g/mol | |
| Sodium persulfate |
| 500 g | |
| Na2S2O8 | |
| UN1505 | |
| CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
| disodium;sulfonatooxy sulfate | |
| 62655 | |
| ≥99% (RT) |
Safety and Handling
P210-P220-P261-P280-P284-P370 + P378
RTECSNumber : SE0525000