missing translation for 'onlineSavingsMsg'
Learn More

Sodium perrhenate, 99+% (metals basis), Re 68%

Catalog No. AA1141204
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1141204 2 g
AA1141209 10 g
AA1141218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1141204 Supplier Thermo Scientific Chemicals Supplier No. 01141204
Only null left

CAS: 13472-33-8 | NaO4Re | 273.193 g/mol

Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13472-33-8
Molecular Formula NaO4Re
Molecular Weight (g/mol) 273.193
MDL Number MFCD00014231
InChI Key KMISVFTVBOPTAI-UHFFFAOYSA-N
Synonym sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium
PubChem CID 5107658
IUPAC Name sodium;oxido(trioxo)rhenium
SMILES [O-][Re](=O)(=O)=O.[Na+]

Specifications

Melting Point 300°C
Color Yellow
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99+% (metals basis)
Solubility Information Soluble in water and ethanol.
Formula Weight 273.19
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 5.39
Chemical Name or Material Sodium perrhenate, Re 68%
Show More Show Less
Hazard Category H272-H315-H319
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 236-742-9
RTECSNumber WD3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only