missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Perchlorate Monohydrate, purum p.a., ≥98% (AT), Honeywell™
Purum p.a., crystallized,≥98.0% (T)
$178.83 - $178.83
Chemical Identifiers
| CAS | 7-3-7791 |
|---|---|
| Molecular Formula | ClH2NaO5 |
| Molecular Weight (g/mol) | 140.45 |
| MDL Number | MFCD00149164 |
| InChI Key | IXGNPUSUVRTQGW-UHFFFAOYSA-M |
| Synonym | sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate |
| PubChem CID | 516899 |
| IUPAC Name | sodium;perchlorate;hydrate |
| SMILES | O.[O-]Cl(=O)(=O)=O.[Na+] |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
| 7-3-7791 | |
| 140.45 | |
| IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
| 516899 | |
| O.[O-]Cl(=O)(=O)=O.[Na+] |
| ClH2NaO5 | |
| MFCD00149164 | |
| sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
| sodium;perchlorate;hydrate |
Specifications
| 7-3-7791 | |
| ClH2NaO5 | |
| MFCD00149164 | |
| sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
| O.[O-]Cl(=O)(=O)=O.[Na+] | |
| 140.45 | |
| 140.46g/mol | |
| Sodium perchlorate monohydrate |
| 250 g | |
| NaClO4 · H2O | |
| UN1502 | |
| IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
| sodium;perchlorate;hydrate | |
| 516899 | |
| ≥98% (T) |
Safety and Handling
P220