missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, Monohydrate, ACS, 85-90%, Spectrum™ Chemical

Catalog No. 18613769
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613769 25 g Amber Glass Bottle
18613770 125 g Amber Glass Bottle
18613771 500 g Amber Glass Bottle
18613772 2.5 kg Amber Glass Bottle
18613773 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613769 Supplier Spectrum Chemical Mfg Cor Supplier No. S137825GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7-3-7791

Specifications

pH 6.0 to 8.0
CAS Max % 1
Quantity 25 g
Percent Purity 85 to 90%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.