missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate monohydrate, ACS, 85.0-90.0%

Catalog No. AA1162230
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1162230 250 g
AA11622A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1162230 Supplier Thermo Scientific Chemicals Supplier No. 01162230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7791-07-3 | ClH2NaO5 | 140.45 g/mol

Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7-3-7791
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 130°C (decomposition)
Color White
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 85.0-90.0%
Solubility Information Soluble in water.
Formula Weight 140.48 (122.46anhydrous)
Grade ACS
Sensitivity Hygroscopic
Appearance White crystals
Density 2.02
Chemical Name or Material odium perchlorate monohydrate, For analysis ACS
Show More Show Less
Hazard Category H271-H302-H319-H373
Precautionary Statement P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P305+P351+P338-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 231-511-9
RTECSNumber SC9850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only