missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate Monohydrate, 98%, ACS Reagent, Honeywell Fluka™

ACS Reagent, 98%

$471.70 - $471.70

Chemical Identifiers

CAS 7-3-7791
ChEBI CHEBI:78672
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045890
View Documents
Honeywell-Fluka
310514500G
500 g Poly Bottle
Each for $471.70
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7-3-7791
CHEBI:78672

Specifications

469°C
500 g
NaClO4 · H2O
UN1502
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+);chloride
62683
140.46g/mol
ACS Reagent
2.02g/cm3
7-3-7791
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
[Cl-].[Rb+]
120.92
CHEBI:78672
98%
Poly Bottle
Sodium Perchlorate Monohydrate
Videos
Safety and Handling

Safety and Handling

P220

H271-H302

DOTInformation : 5.1/PGII

EINECSNumber : 231-511-9

RTECSNumber : SC9850000

SDS
missing translation for 'documents'

missing translation for 'documents'