missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate monohydrate, 85+%, ACS reagent

Catalog No. p-4510594
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC343631000 100 g Plastic bottle
AC343630010 1 kg Plastic bottle
AC343630025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC343631000 Supplier Thermo Scientific Chemicals Supplier No. 343631000
Only null left

CAS: 7791-07-3 | ClNaO4·H2O | 140.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7-3-7791
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 130.0°C
Color White to Yellow
pH 6.0 to 8.0 (5% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 85.0 to 90% (as NaClO4) (Argentometry)
Molecular Formula ClNaO4·H2O
Linear Formula ClNaO4·H2O
Merck Index 15,8784
Solubility Information Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 140.46
Formula Weight 140.46
Percent Purity ≥85%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium perchlorate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.

RUO – Research Use Only