missing translation for 'onlineSavingsMsg'
Learn More

Sodium percarbonate, 13-14% active oxygen

Catalog No. AAA1604522
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1604522 100 g
AAA1604536 500 g
AAA160450E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1604522 Supplier Thermo Scientific Chemicals Supplier No. A1604522

CAS: 15630-89-4 | Na2CO3·1.5H2O2

Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong reducing agents, strong acids, organic materials and powdered metals.

Chemical Identifiers

CAS 15630-89-4
Molecular Formula Na2CO3·1.5H2O2
MDL Number MFCD00043204
Synonym Hydrogen peroxide-Sodium carbonate adduct

Specifications

Quantity 100 g
UN Number UN1479
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 157.01
Chemical Name or Material Sodium percarbonate, 13 to 14% active oxygen
Hazard Category H272-H302-H318
Hazard Statement GHS H Statement
H272-H318
May intensify fire; oxidizer.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 239-707-6
RTECSNumber FG0750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only