missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, Reagent Grade

Catalog No. AA3338314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3338314 25 g
AA3338322 100 g
AA3338336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3338314 Supplier Thermo Scientific Chemicals Supplier No. 03338314
Only null left

CAS: 10486-00-7 | NaBO3·4H2O

Sodium perborate tetrahydrate is mainly used in textile mills as a bleaching agent for a variety of fabrics, namely, wool, cotton, linen, rayon, etc, and as oxidizing and fixing agent for dyes. It is also used in dentifrices, denture cleaners and pharmaceuticals due to its cleansing, deodorizing and antiseptic properties and is incorporated in soaps and detergent powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 10486-00-7
MDL Number MFCD00149231

Specifications

Melting Point 60°C (decomposition)
Physical Form Granules
Quantity 25 g
Molecular Formula NaBO3·4H2O
Solubility Information Soluble in water (23g/L) at 20°C.
Formula Weight 153.86 (81.84 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Density 1.73
Chemical Name or Material Sodium Perborate Tetrahydrate
Hazard Category H318-H332-H335-H360Df
Precautionary Statement P201-P202-P261-P271-P281-P304+P340-P305+P351+P338-P308+P313-P310-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 239-172-9
RTECSNumber SC7350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only