missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perborate Tetrahydrate 97.0+%, TCI America™

Catalog No. p-7135980
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S088725G 25 g
S0887500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S088725G Supplier TCI America Supplier No. S088725G
Only null left

Chemical Identifiers

CAS 10486-00-7
Molecular Formula BNaO3
MDL Number MFCD00149231

Specifications

Melting Point 60°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 1479
Formula Weight 81.80
Percent Purity ≥97.0% (T)
Chemical Name or Material Sodium Perborate Tetrahydrate [for Biochemical Research]
EINECSNumber (1)-0826
RTECSNumber SC7350000
TSCA No