missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, 97%

Catalog No. p-7080605
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1018922 100 g
AAA1018936 500 g
AAA101890E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1018922 Supplier Thermo Scientific Chemicals Supplier No. A1018922
Only null left

CAS: 10486-00-7 | NaBO3·4H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

Solubility
Soluble in water.

Notes
Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.

Chemical Identifiers

CAS 10486-00-7
Molecular Formula NaBO3·4H2O
MDL Number MFCD00149231

Specifications

Melting Point 60°C (decomposition)
Density 1.73
pH ∼10
Odor Odorless
Quantity 100 g
UN Number UN1479
Sensitivity Hygroscopic
Merck Index 14,8652
Solubility Information Soluble in water.
Formula Weight 153.86 (81.84 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium perborate tetrahydrate
Show More Show Less
Hazard Category H318-H332-H335-H360Df
Hazard Statement GHS H Statement
H272-H360-H318-H302-H335
May intensify fire
oxidizer.
May damage fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P271-P281-P304+P340-P305+P351+P338-P308+P313-P310-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 239-172-9
RTECSNumber SC7350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only