missing translation for 'onlineSavingsMsg'
Learn More

Sodium p-Toluenesulfinate 98.0+%, TCI America™

Catalog No. T027525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T027525G 25 g
T0275500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T027525G Supplier TCI America Supplier No. T027525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 824-79-3
Molecular Formula C7H7NaO2S
Molecular Weight (g/mol) 178.181
MDL Number MFCD00013136
InChI Key KFZUDNZQQCWGKF-UHFFFAOYSA-M
Synonym sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech
PubChem CID 2723791
IUPAC Name sodium;4-methylbenzenesulfinate
SMILES CC1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White-Yellow
Quantity 25 g
Formula Weight 178.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium p-Toluenesulfinate
EINECSNumber (3)-1873
RTECSNumber XT4725000
TSCA Yes