missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390931
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02004181 125 g Wide Mouth Amber Glass Bottle
02004182 500 g Poly Bottle
2 options

Catalog No. 02004181

Supplier: Avantor 380104

Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Physical Form Powder
Quantity 125 g
IUPAC Name disodium oxalate
Formula Weight 134
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Sodium Oxalate, Titrimetry Standard