missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Oxalate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| IUPAC Name | disodium;oxalate |
| Formula Weight | 134 |
| Percent Purity | 99% to 100% |
| Grade | Not Available |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 2.34kg |
| Chemical Name or Material | Sodium Oxalate, Titrimetry Standard |