missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, For ACS analysis, 99.96%, MP Biomedicals™

$112.38 - $112.38

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19143980
View Documents
MP Biomedicals Inc
0219143980
100 g
Each for $112.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62-76-0
134
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
6125
[Na+].[Na+].[O-]C(=O)C([O-])=O
C2Na2O4
MFCD00012465
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
disodium oxalate

Specifications

62-76-0
2.34 (water = 1)
C2Na2O4
100 g
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
disodium oxalate
6125
Crystalline Powder
White
0.9996
MFCD00012465
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
[Na+].[Na+].[O-]C(=O)C([O-])=O
134
134
Sodium oxalate, For ACS analysis
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'