missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, ACS reagent

Catalog No. p-4440164
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446961000 100 g Plastic bottle
AC446965000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446961000 Supplier Thermo Scientific Chemicals Supplier No. 446961000
Only null left

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 250.0°C to 270.0°C
Quantity 100 g
Linear Formula NaO2CCO2Na
Merck Index 15,8781
Solubility Information Solubility in water: 37g/L (20°C). Other solubilities: 6.3 g/100mL water (100°C), insoluble in ethanol and ether
Formula Weight 134
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium oxalate
Hazard Category GHS Signal Word: Warning
EINECSNumber 200-550-3

RUO – Research Use Only