missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCS0720500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0720500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0720500 Supplier Ricca Chemical Company Supplier No. RDCS0720500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging White Poly Wide Mouth Bottle
Quantity 500 g
Percent Purity Min. 99.5%
Grade ACS
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sodium Oxalate
ShelfLife 60 months