missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, ACS Grade, ≥ 99.5%, LabChem™

Catalog No. LC247101
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC247101 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC247101 Supplier LabChem Supplier No. LC247101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Color White
CAS Max % 100
Physical Form Powder
Quantity 500 g
Linear Formula Na2C2O4
Solubility Information Soluble in water
Formula Weight 134
Percent Purity ≥99.5%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2.34g/cm3
Decomposition Information Carbon monoxide; Carbon dioxide
Density 2.34g/cm3
Chemical Name or Material Sodium Oxalate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed or in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 200-550-3
Recommended Storage Room Temperature