Learn More
Sodium oxalate, 99%
CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Supplier: Thermo Scientific Chemicals A116480E
Description
Sodium oxalate is used as reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium oxalate is used as reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.
Solubility
Soluble in water and formic acid. Insoluble in ether and alcohol.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| Sodium oxalate | |
| ∼250°C to 270°C (decomposition) | |
| Odorless | |
| MFCD00012465 | |
| 3631622 | |
| 14,8650 | |
| Soluble in water and formic acid. Insoluble in ether and alcohol. | |
| [Na+].[Na+].[O-]C(=O)C([O-])=O | |
| 134.00 | |
| 134 |
| 62-76-0 | |
| 2.34 | |
| C2Na2O4 | |
| 2500 g | |
| Hygroscopic | |
| sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt | |
| ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
| disodium;oxalate | |
| 6125 | |
| 99% |
Chemical Identifiers
| 62-76-0 | |
| 134.00 | |
| ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
| 6125 | |
| [Na+].[Na+].[O-]C(=O)C([O-])=O |
| C2Na2O4 | |
| MFCD00012465 | |
| sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt | |
| disodium;oxalate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312-H320
EINECSNumber : 200-550-3
RTECSNumber : KI1750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only