missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, ≥99.5%, ACS Reagent, Honeywell Fluka™

Catalog No. 60045356
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045356 100 g Poly Bottle
60045357 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045356 Supplier Honeywell Chemicals Supplier No. 223433100G
Only null left

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Quantity 100 g
Linear Formula NaOOCCOONa
UN Number NONH for all modes of transport
Beilstein 3631622
Formula Weight 134g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Oxalate
Hazard Statement P280
Precautionary Statement H302 + H312
EINECSNumber 200-550-3