missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, 0.100 M (M/10), 0.200 N (N/5), Ricca Chemical

Catalog No. R75000001A
Change view
Click to view available options
Package: Natural Poly; Size: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R75000001A Package: Natural Poly; Size: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R75000001A Supplier Ricca Chemical Company Supplier No. R75000001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 62-76-0, 7732-18-5
Assay Percent Range 0.1000 ± 0.0001 M at 20°C
Molecular Formula C2Na2O4
MDL Number MFCD00012465
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
IUPAC Name disodium oxalate
Molecular Weight (g/mol) 134.00
PubChem CID 6125
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Chemical Name or Material Sodium Oxalate, 0.100 Molar (M/10) (0.200 Normal)
Show More Show Less