Learn More
Sodium orthovanadate, 100mM aq. soln.
CAS: 13721-39-6 | Na3O4V | 183.907 g/mol
$162.93 - $264.79
Chemical Identifiers
| CAS | 13721-39-6 |
|---|---|
| Molecular Formula | Na3O4V |
| Molecular Weight (g/mol) | 183.907 |
| MDL Number | MFCD00003511 |
| InChI Key | IHIXIJGXTJIKRB-UHFFFAOYSA-N |
| Synonym | sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt |
| PubChem CID | 61671 |
| ChEBI | CHEBI:35607 |
| IUPAC Name | trisodium;trioxido(oxo)vanadium |
| SMILES | [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60191AD
|
Thermo Scientific Chemicals
J60191AD |
50 mL |
Each for $162.93
|
|
|||||
|
AAJ60191AE
|
Thermo Scientific Chemicals
J60191AE |
100 mL |
Each for $264.79
|
|
|||||
Description
Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.
Solubility
It is soluble in water (50 mg/ml, )
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
Chemical Identifiers
| 13721-39-6 | |
| 183.907 | |
| IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
| 61671 | |
| trisodium;trioxido(oxo)vanadium |
| Na3O4V | |
| MFCD00003511 | |
| sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt | |
| CHEBI:35607 | |
| [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
Specifications
| Colorless | |
| Liquid | |
| Na3O4V | |
| MFCD00003511 | |
| It is soluble in water (50mg/ml,) | |
| [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] | |
| 183.907 | |
| CHEBI:35607 | |
| 100mM aq. soln. | |
| Sodium orthovanadate |
| 13721-39-6 | |
| 50 mL | |
| Na3VO4 | |
| sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt | |
| IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
| trisodium;trioxido(oxo)vanadium | |
| 61671 | |
| 183.94 | |
| Odorless |
Safety and Handling
H335
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only