missing translation for 'onlineSavingsMsg'
Learn More

Sodium Octanohydroxamate Hydrate 98.0+%, TCI America™

Catalog No. C055725G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C055725G 25 g
C05571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C055725G Supplier TCI America Supplier No. C055725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50292-00-7
Molecular Formula C8H18NNaO3
Molecular Weight (g/mol) 199.226
MDL Number MFCD00058968
InChI Key FYYOBOYZQQPVMW-UHFFFAOYSA-N
Synonym Octanohydroxamic Acid Sodium Salt
PubChem CID 56924515
IUPAC Name sodium;N-oxidooctanamide;hydrate
SMILES CCCCCCCC(=O)N[O-].O.[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 181.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Octanohydroxamate Hydrate